EP Scientific EPA Vials, Borosilicate Glass, with PTFE/Silicone-Lined Cap, Thermo Scientific
Supplier: Thermo Scientific
139-20A 139-20A/CT 139-20C 139-20C/CT 139-20A/EP 139-20C/EP 140-40C 140-40C/CT 140-40C/DB 140-40C/EP 140-40C/TS 140-60C 141-40A 141-40A/CT 141-40A/DB 141-40A/EP 141-40A/TS 339-20A 339-20C 340-40C 340-40C/DB 340-60C 341-40A 341-40A/DB 341-60A
EP139-20ACS 590.13 CAD
EP139-20A EP139-20ACT EP139-20C EP139-20CCT EP13920AEP EP13920CEP EP140-40C EP140-40CCT EP140-40CDB EP140-40CEP EP140-40CTS EP140-60C EP141-40A EP141-40ACT EP141-40ADB EP141-40AEP EP141-40ATS EP339-20A EP339-20C EP340-40C EP340-40CDB EP340-60C EP341-40A EP341-40ADB EP341-60A
EP Scientific EPA Vials, Borosilicate Glass, with PTFE/Silicone-Lined Cap, Thermo Scientific
Vials
Vials processed according to Level 1 are certified to meet or exceed EPA standards for volatiles.
Vials processed according to Level 3 are not pre-cleaned and are ready for your own cleaning procedure. Level 3 vials are assembled and meet EPA recommended guidelines for sample container material component specifications.
Each case is custody-sealed. Each vial is lot number labeled for traceability to the enclosed Certificate of Analysis.
Ordering information: Vials are supplied with open-top or closed-top polypropylene caps with 3 mm (0.125") or 1.5 mm (0.060") PTFE/silicone septa.More Product Information