Methyl acetoacetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.00107.0100 8.00107.1000
EM8.00107.0100CS 711 CAD
EM8.00107.0100 EM8.00107.1000 CA8.00107.0100 CA8.00107.1000
Methyl acetoacetate for synthesis, Sigma-Aldrich®
Methyl acetoacetate
Formula: C₅H₈O₃
Boiling Pt: 170 °C (1013 hPa)
Melting Pt: –39 °C
Density: 1.076 g/cm³ (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00008784
CAS Number: 105-45-3
EINECS: 203-299-8
Merck Index: 12,06090

Order Now

Specification Test Results

Solubility 863 g/l (20 °C)
Melting Point -39 °C
Molar Mass 116.12 g/mol
Boiling Point 170 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 67 °C
Refractive Index 1.4185 (20 °C, 589 nm)
Explosion Limit 1.3 - 14.2 %(V)
Density 1.08 g/cm3 (20 °C)
pH 3.8 (863 g/l, H2O, 20 °C)
Ignition Point 260 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR