Potassium chloride, Supelco®

Supplier: MilliporeSigma

Synonyms: Potassiumchloride

M-Clarity™ quality level = MQ300

1.04933.9025
EM1.04933.9025EA 18312.2 CAD
EM1.04933.9025
Potassium chloride, Supelco®
Potassium chloride
Formula: KCl
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
MDL Number: MFCD00011360
CAS Number: 7447-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR