Bismuth(III) oxide ≥99.0% (by complexometric analysis), extra pure, Supelco®

Supplier: MilliporeSigma

Synonyms: Bismuth Trioxide, Dibismuth trioxide

1.01862.9025 1.01862.9025 1.01862.1000
EM1.01862.9025EA 19800.56 CAD
EM1.01862.9025 CA1.01862.9025 EM1.01862.1000
Bismuth(III) oxide ≥99.0% (by complexometric analysis), extra pure, Supelco®
Bismuth (III) oxide
Formula: BiO₃
MW: 465.96 g/mol
Boiling Pt: 1890
Melting Pt: 825
Density: 8.93 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Order Now

Specification Test Results

Melting Point 820 °C
Molar Mass 465.95 g/mol
Bulk Density 1000 kg/m3
Boiling Point 1890 °C
Density 8.93 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR