Epoxy Resin Work Surfaces, Durcon Laboratory Tops
Supplier: Durcon
DURCON04961 DURCON24961 DURCON31961
DUDURCON04961EA 792.18 CAD
DUDURCON04961 DUDURCON24961 DUDURCON31961
Epoxy Resin Work Surfaces, Durcon Laboratory Tops
Furniture Tables and Workbenches
These black, molded epoxy resin worktops are solid, durable, and nonporous.
  • Thickness: 2.5 cm (1")
  • Work surfaces feature a standard beveled edge finish
  • Fabricated to exact specifications
They feature excellent physical, chemical, stain, and heat resistance, and will not delaminate, swell, or spall. Work surfaces have a smooth, low-glare exterior with a standard drip groove.
Ordering information: Work surfaces with a radius edge finish, and flat epoxy resin sheets of up to 167.6W×254L cm (66×100") are also available; Black matte resin backsplashesMore Product Information