VWR® Contour™ Standing Height Sink Cabinets
Supplier: VWR International
CFG-2004-A CFG-2004-L CFG-2005-A CFG-2006-A CFG-2008-A CFG-2009-A CFG-20452A CFG-20458A CFG-2006-P CWG-2004-A CWG-2005-A CWG-2006-A CWG-2008-A CWG-2009-A CWG-20452A CWG-20458A CWG-2004-L
CFG-2004-AEA 2414.35 CAD
CFG-2004-A CFG-2004-L CFG-2005-A CFG-2006-A CFG-2008-A CFG-2009-A CFG-20452A CFG-20458A CFG-2006-P CWG-2004-A CWG-2005-A CWG-2006-A CWG-2008-A CWG-2009-A CWG-20452A CWG-20458A CWG-2004-L
VWR® Contour™ Standing Height Sink Cabinets
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
A 2.54 cm (1") thick work surface creates an overall height of 91.4 cm (36"). All cupboard cabinets have one full-width adjustable shelf. Sink cabinets with center bench end comes furnished with finished back panels that may be removed when not required. Finished back adds 2.54 cm (1") to depth, for an overall cabinetMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable