VWR® Contour™ Mobile Cabinets, Avantor
Supplier: Kewaunee
CFD1812B41-INT CFD1814B41-INT CFD-1842-B41 CFD-1844-B41 CFE-1402-B41 CFE-1402L-B41 CFE-1404-B41 CFE-1404L-B41 CFE-1405-B41 CFE-1416-B41 CWD1812B41-INT CWD1814B41-INT CWD1842B41-INT CWD1844B41-INT CWE-1402-B41 CWE-1402L-B41 CWE-1404-B41 CWE-1404L-B41 CWE-1405-B41 CWE-1416-B41 CWC1032B41-INT CWC1032LB41INT CWC1034B41-INT CWC1034LB41INT CWC1035B41-INT CWC1036B41-INT CWD-1002-B41 CWD-1004-B41 CWD-1005-B41 CWD-1006-B41 CFC1032B41-INT CFC-1032L-B41 CFC1034B41-INT CFC-1034L-B41 CFC1035B41-INT CFC1036B41-INT CFD-1002-B41 CFD-1004-B41 CFD-1005-B41 CFD-1006-B41
CFD-1812-B41EA 2531.08 CAD
CFD-1812-B41 CFD-1814-B41 CFD-1842-B41 CFD-1844-B41 CFE-1402-B41 CFE-1402L-B41 CFE-1404-B41 CFE-1404L-B41 CFE-1405-B41 CFE-1416-B41 CWD-1812-B41 CWD-1814-B41 CWD-1842-B41 CWD-1844-B41 CWE-1402-B41 CWE-1402L-B41 CWE-1404-B41 CWE-1404L-B41 CWE-1405-B41 CWE-1416-B41 CWC-1032-B41 CWC-1032L-B41 CWC-1034-B41 CWC-1034L-B41 CWC-1035-B41 CWC-1036-B41 CWD-1002-B41 CWD-1004-B41 CWD-1005-B41 CWD-1006-B41 CFC-1032-B41 CFC-1032L-B41 CFC-1034-B41 CFC-1034L-B41 CFC-1035-B41 CFC-1036-B41 CFD-1002-B41 CFD-1004-B41 CFD-1005-B41 CFD-1006-B41
VWR® Contour™ Mobile Cabinets, Avantor
Furniture Storage Cabinets
American-made, steel mobile cabinets, shipped fully assembled.
  • Numerous sizes and configurations
  • Two color options
  • Ships fully assembled
  • 3" Swivel locking caster
  • Powder-coated
The VWR® Contour™ mobile cabinet line is designed to integrate seamlessly into your existing Contour™ lab design, with the integrated flush pull design. Optioned in both light neutral and snow white powder coat, this cold rolled steel units come fully assembled and ready for use right out of the box.
Certifications:
Complies with SEFA 10 requirements.
Delivery information:More Product Information