VWR® Contour™ ADA Height Base Cabinets, Acid Storage
Supplier: VWR International
CFG-3804-A CFG-3804-L CFG-3805-A CFG-3806-A CFG-3808-A CWG-3804-A CWG-3804-L CWG-3805-A CWG-3806-A CWG-3808-A
CFG-3804-AEA 4097.11 CAD
CFG-3804-A CFG-3804-L CFG-3805-A CFG-3806-A CFG-3808-A CWG-3804-A CWG-3804-L CWG-3805-A CWG-3806-A CWG-3808-A
VWR® Contour™ ADA Height Base Cabinets, Acid Storage
Furniture Casework
Heavy-gauge steel full-frame construction, specifically designed for the storage of corrosive chemicals.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
  • Lined with a molded one piece linear LDPE tub with coved corners and a 1" lip with 18" PE sheet
  • Doors are latched using a nylon roller catch
Each cabinet is furnished with a PVC vent pipe for venting to the fume hood above
A 2.54 cm (1") thick workMore Product Information