VWR® Contour™ Suspended Cabinets
Supplier: VWR International
CFC-4032-A CFC-4032-B CFC-4032-BL CFC-5032-B CFC-5032-BL CFD-4002-A CFD-4812-A CFD-4852-B CFD-5002-B CFD-5852-B CFE-4402-A CFE-4402-B CFE-4402-BL CFE-4402-L CFC-5032-A CFC-5032-L CFC-4032-L CWC-5032-A CWC-5032-L CWD-4002-A CWD-4852-B CWE-4402-A CWE-4402-L CWD-4812-A CWE-4402-B
CFC-4032-AEA 2319.28 CAD
CFC-4032-A CFC-4032-B CFC-4032-BL CFC-5032-B CFC-5032-BL CFD-4002-A CFD-4812-A CFD-4852-B CFD-5002-B CFD-5852-B CFE-4402-A CFE-4402-B CFE-4402-BL CFE-4402-L CFC-5032-A CFC-5032-L CFC-4032-L CWC-5032-A CWC-5032-L CWD-4002-A CWD-4852-B CWE-4402-A CWE-4402-L CWD-4812-A CWE-4402-B
VWR® Contour™ Suspended Cabinets
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Cabinets are designed to hang beneath table frames. Light neutral color.
Order Now
SPECIFICATIONS
Environmentally Preferable