VWR® Contour™ Apron Rails
Supplier: VWR International
CFA060424-A CFA060430-A CFA060436-A CFA060448-A CFA060460-A CFA060472-A CFA560436-A CFA560448-A CFA560460-A CFA560472-A CWA060424-A CWA060430-A CWA060436-A CWA060460-A CWA060472-A CWA060448-A
CFA060424-AEA 283.63 CAD
CFA060424-A CFA060430-A CFA060436-A CFA060448-A CFA060460-A CFA060472-A CFA560436-A CFA560448-A CFA560460-A CFA560472-A CWA060424-A CWA060430-A CWA060436-A CWA060460-A CWA060472-A CWA060448-A
VWR® Contour™ Apron Rails
Furniture Casework
These rails are only to be used between base cabinets.
  • Contour casework is manufactured of prime grade, cold-rolled, U.S. Standard sheet steel
  • No ragged edges
  • Light neutral color
The complete apron is used to create a knee space for a desk area or to create a free-standing table. Apron components are sold separately.
Order Now
SPECIFICATIONS
Environmentally Preferable