EVERHUTCH NESTCART Mobile Utility Carts, Kewaunee
Supplier: Kewaunee
MCB00D3018M-00 MCB00D3018M-01 MCB00D3024M-00 MCB00D3024M-01 MCB00D3030M-00 MCB00D3030M-01 MCB00D3318M-00 MCB00D3318M-01 MCB00D3324M-00 MCB00D3324M-01 MCB00D3618M-00 MCB00D3618M-01 MCB00D3624M-00 MCB00D3624M-01 MCB00D3630M-00 MCB00D3630M-01 MCB03C3624LM0 MCB03C3624LM1 MCB03C3624RM0 MCB03C3624R1 MCB08D3618M-00 MCB08D3618M-01 MCB08D3624M-00 MCB08D3624M-01 MCB26D3018M-00 MCB26D3018M-01 MCB30D3018M-00 MCB30D3018M-01 MCB30D3024M-00 MCB30D3024M-01 MCB30D3030M-00 MCB30D3030M-01 MCB30D3318M-00 MCB30D3318M-01 MCB30D3324M-00 MCB30D3324M-01 MCB30D3630M-00 MCB32D3330M-00 MCB32D3330M-01 MCB40E3024LM0 MCB40E3024LM1 MCB50D3318M-00 MCB50D3318M-01 MCB50D3324M-00 MCB50D3324M-01 MCB50D3618M-00 MCB50D3618M-01 MCB50D3624M-00 MCB50D3624M-01 MCB51D3630M-00 MCB51D3630M-01 MCB52D3630M-00 MCB52D3630M-01
CB00D301800EA 3137.07 CAD
CB00D301800 CB00D301801 CB00D302400 CB00D302401 CB00D303000 CB00D303001 CB00D331800 CB00D331801 CB00D332400 CB00D332401 CB00D361800 CB00D361801 CB00D362400 CB00D362401 CB00D363000 CB00D363001 CB03C3624L00 CB03C3624L01 CB03C3624R00 CB03C3624R01 CB08D361800 CB08D361801 CB08D362400 CB08D362401 CB26D301800 CB26D301801 CB30D301800 CB30D301801 CB30D302400 CB30D302401 CB30D303000 CB30D303001 CB30D331800 CB30D331801 CB30D332400 CB30D332401 CB30D363000 CB32D333000 CB32D333001 CB40E3024L00 CB40E3024L01 CB50D331800 CB50D331801 CB50D332400 CB50D332401 CB50D361800 CB50D361801 CB50D362400 CB50D362401 CB51D363000 CB51D363001 CB52D363000 CB52D363001
EVERHUTCH NESTCART Mobile Utility Carts, Kewaunee
Trolleys
Mobile units with drawer configurations for storage in clinical and work areas.
  • Mobile cart on wheels
  • Sitting, ADA, and standing height cabinets
  • Drawer configurations
  • Under counter storage
  • File cabinet
  • Supply storage
Ideal for supply storage in clinical areas such as patient rooms. Provides added storage space under a counter to maximize the space. Variable heights allow for sitting, ADA, and standing height work surfaces (30, 33, 36") with full extension drawers. The cart is on four swivel locking casters for easy movement.
Ordering information: More Product Information