Triethylamine ≥99.5% for HPLC, Supelco®

Supplier: MilliporeSigma
Danger

TX1202-5
CATX1202-5EA 500 CAD
CATX1202-5
Triethylamine ≥99.5% for HPLC, Supelco®
Triethylamine

Triethylamine. Suitable for multiple applications, these reagents meet or exceed ACS specifications.

Formula: (C₂H₅)₃N
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.729 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 (8),II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% min
Appearance Clear, free of paticulate matter
Color (APHA) 20 max.
Identity (IR-spectrum) Conforms
UV Abs. (0.1% water, 254 nm) 0.10AU max.
UV Abs. (0.1% water, 280 nm) 0.04AU max.
UV Abs. (0.1% water, 300 nm) 0.02AU max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR