Triethylamine ≥99.5%, Supelco®

Supplier: MilliporeSigma
Danger

TX1200-11 TX1200-2 TX1200-5
CATX1200-11EA 261 CAD
CATX1200-11 CATX1200-2 CATX1200-5
Triethylamine ≥99.5%, Supelco®
Triethylamine
Formula: (C₂H₅)₃N
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.729 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 (8),II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% min
Appearance Clear, free of particulate matter
Color (APHA) 15 max
Identity (IR-spectrum) Conforms
Water (H2O) 0.1% max

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR