BL21 Competent Cells, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
230280 200131 200132 200133 230130 230132 230134 230240 230245 230255
CA76193-370PK 383.25 CAD
CA76193-370 CA97066-330 CA99900-860 CAST200131 CAST200132 CAST200133 CAST230130 CAST230132 CAST230134 CAST230240 CAST230245 CAST230255 CAST230275
BL21 Competent Cells, Agilent Technologies
Competent Cells
BL21 competent cells are an all-purpose strain for high-level protein expression and easy induction.
  • High-level protein expression and easy induction
  • Can be used with non-T7 RNA polymerase protein expression systems
  • Allows the tightest control of protein expression for extremely toxic proteins
  • Suitable for expressing proteins from all species, including mammals
  • Ideal for difficult protein expression, especially when codon bias is a problem
  • Increases transformation efficiency with Hte phenotype
  • Derived from Agilent’s high performance BL21-Gold competent
More Product Information