Anti-TYMS Sheep Polyclonal Antibody

Supplier: Rockland Immunochemical
200-601-199S
CARL200601199SEA 229.01 CAD
CARL200601199S
Anti-TYMS Sheep Polyclonal Antibody
Antibodies
THYMIDYLATE SYNTHASE antibody has been tested for use in ELISA, immunoprecipitation, immunofluorescence microscopy, immunohistochemistry and immunoblotting.  The antibody recognizes the expected additional band corresponding to the ternary complex of hTS-

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500 - 1:2000; Other Dilution: User Optimized

Type: Primary
Antigen: TYMS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR