Washed Pooled Cells, Rockland Immunochemicals
Supplier: Rockland Immunochemical
R401-0050 R401-0100 R402-0050 R402-0100 R403-0050 R403-0100 R404-0050 R404-0100 R405-0100 R406-0050 R406-0100 R407-0050 R407-0100
CAR401-0050EA 390.01 CAD
CAR401-0050 CAR401-0100 CAR402-0050 CAR402-0100 CAR403-0050 CAR403-0100 CAR404-0050 CAR404-0100 CAR405-0050 CAR405-0100 CAR406-0050 CAR406-0100 CAR407-0050 CAR407-0100
Washed Pooled Cells, Rockland Immunochemicals
Blood
Suspensions are suitable for complement fixation, hemagglutination, hemadsorption, and other related serological procedures
All suspensions are 10% cell solutions in saline, except for RLR406-0050 and RLR406-0100. These two suspensions are 100% cells. Suspensions contain no stabilizers or preservatives.
Order Now
SPECIFICATIONS
Environmentally Preferable