Animal Whole Blood, Rockland Immunochemicals
Supplier: Rockland Immunochemical
R105-0050 R105-0100 R107-0100 R109-0050 R109-0100 R111-0050 R111-0100 R300-0050 R302-0050 R302-0100 R305-0050 R305-0100 R306-0050 R307-0050 R307-0100 R308-0050 R309-0050 R309-0100 R310-0050 R310-0100 R311-0050 R311-0100
CAR105-0050EA 978 CAD
CAR105-0050 CAR105-0100 CAR107-0100 CAR109-0050 CAR109-0100 CAR111-0050 CAR111-0100 CAR300-0050 CAR302-0050 CAR302-0100 CAR305-0050 CAR305-0100 CAR306-0050 CAR307-0050 CAR307-0100 CAR308-0050 CAR309-0050 CAR309-0100 CAR310-0050 CAR310-0100 CAR311-0050 CAR311-0100
Animal Whole Blood, Rockland Immunochemicals
Blood
Use for DNA isolation
Blood is obtained from healthy, non-medicated animals. All blood, except hamster, mouse, pigeon, and rat blood is sterile.
Ordering information: For additional coagulants, sizes, or species, contact your Avantor representative.
Order Now
SPECIFICATIONS
Environmentally Preferable