Poly-HRP Dilution Buffer, Thermo Scientific

Supplier: Thermo Scientific

N500
CAPIN500EA 279 CAD
CAPIN500
Poly-HRP Dilution Buffer, Thermo Scientific
Buffers Assay Buffers

Thermo Scientific Pierce Poly-HRP Dilution Buffer is optimized to ensure optimal performance with Poly-HRP Streptavidin.


Features of Poly-HRP Dilution Buffer:


Formulation: 1% biotin-free casein in a PBS buffer solution.
Application: Dilution of Poly-HRP Streptavidin (Product # N200): Gently invert bottle and swirl to mix before use. Do not vortex. Use at full strength. Refer to Poly-HRP Streptavidin Certificate of Analysis for recommended dilution ranges. Dilute Poly-HRP Streptavidin immediately before use.


Formulation: 1% biotin-free casein in a PBS buffer solution.


For Research Use Only. Not for use in diagnostic procedures.


Ordering information: Shipped on cold packs. Store at 2-80˚C immediately upon receipt.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR