Pierce™ Affinity Chromatography Cartridges, Protein G, Thermo Scientific
Supplier: Thermo Scientific
89926 89927
CAPI89926EA 616.11 CAD
CAPI89926 CAPI89927
Pierce™ Affinity Chromatography Cartridges, Protein G, Thermo Scientific
Chromatography Columns
These cartridges are convenient, ready-to-use prepacked devices for isolation and purification of mammalian polyclonal and monoclonal antibodies from serum, ascites and cell culture supernatants.
  • Protein G: Immobilized Protein G is ideal for polyclonal IgG purification from mouse, human, cow, goat and sheep serum, including human IgG3 and mouse IgG1 isotypes
  • Agarose resin: Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Inert and stable: Superior manufacturing method immobilizes Protein G
More Product Information