Pierce™ Centrifuge Columns for Affinity Chromatography, Thermo Scientific
Supplier: Thermo Scientific
89869 89868 89896 89897 89898
CAPI-89869CS 629.67 CAD
CAPI-89869 CAPI89868 CAPI89896 CAPI89897 CAPI89898
Pierce™ Centrifuge Columns for Affinity Chromatography, Thermo Scientific
Filters Centrifugal Filters
Pierce™ Centrifuge Columns are convenient tools for handling 40 µl to 10 ml of an affinity support for purification.
  • Made from low protein binding polypropylene for compatibility with protein purification
  • Filter type, pore size: Polyethylene filter, ~30 µm pore size
  • Twist-off bottom
These centrifuge columns can be used in either spin or gravity-flow format.
Applications:
Affinity purification, affinity chromatography
Immunodepletion
Ion exchange resins
Gel filtration, desalting, buffer exchange
The spin format reduces sample processing time andMore Product Information