Pierce™ 1- Step Human High-Yield Maxi IVT Kit, Thermo Scientific
Supplier: Thermo Scientific
88892
CAPI88892EA 14940.12 CAD
CAPI88892
Pierce™ 1- Step Human High-Yield Maxi IVT Kit, Thermo Scientific
Protein Expression Protein Expression Systems
The Thermo Scientific 1-Step human high-yield In Vitro Translation (IVT) Kits enable the expression of functional proteins with 10 to 100 times greater yield per ml than other mammalian IVT systems.
  • High expression with up to 750 µg/ml of expressed protein
  • Low variability between experiments
  • Express high levels of protein with 6 hours to overnight incubation
  • Obtain functionally active proteins, including those containing disulfide bonds
  • Transcription and translation is performed in one reaction step
  • Amenable to incorporation of heavy or unnatural amino acids
The 1-Step High-Yield IVT System uses modified HeLa cell extracts to take advantage of the robust human translation machinery and generate functional full-length proteins. In this system, protein expression is performed in a proprietary dialysis device that allows a continuous supply of nucleotides, amino acids and energy generating substrates into the reaction while removing inhibitors of proteins synthesis. This continuous-exchange cell-free (CECF) system enables protein expression in an overnight incubation of up to 750 µg/ml. The complete mini-scale kit includes all the components required for transcription and translation of a recombinant gene, including an optimised expression vector.
Delivery information: Each 1- Step Human High-Yield Maxi IVT Kit contains HeLa lysate (4 x 500 µl), accessory proteins (4 x 100 µl), reaction mix (4 x 200 µl), 5X dialysis buffer (23 ml), positive control DNA (pCFE-GFP - 10 µg), cloning vector (pT7CFE1-NHis-GST-CHA - 10 µg), microdialysis devices (2), and nuclease-free water (3 x 50 ml).