Pierce™ Protein Magnetic Beads, Protein A, Protein G, Thermo Scientific
Supplier: Thermo Scientific
88845 88846 88847 88848
CAPI88845EA 590.92 CAD
CAPI88845 CAPI88846 CAPI88847 CAPI88848
Pierce™ Protein Magnetic Beads, Protein A, Protein G, Thermo Scientific
Beads Magnetic Beads
Thermo Scientific Pierce protein magnetic beads are high-capacity and high-throughput affinity particles for antibody purification and immunoprecipitation methods using manual or robotic magnetic separators.
  • Higher efficiency in yield of IP target antigens
  • Low non-specific binding of stable, pre-blocked beads provide highly purified product
  • Magnetic beads eliminate resin loss and provide for more efficient separation than traditional IP methods that use only centrifugation
  • Beads are compatible with manual and automated workflows making it versatile