L(+)-Lysine dihydrochloride, lyophilised powder for SILAC

Supplier: Thermo Scientific

Synonyms: H-Lys-OH·2HCl, H-Lys-OH.2HCl, (S)-(+)-Lysine dihydrochloride, (S)-(+)-2,6-Diaminohexanoic acid dihydrochloride, (S)-(+)-2,6-Diaminocaproic acid dihydrochloride

88429
CAPI88429EA 417.52 CAD
CAPI88429
L(+)-Lysine dihydrochloride, lyophilised powder for SILAC
L(+)-Lysine dihydrochloride

Thermo Scientific Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits.


  • Efficient—100% Label Incorporation into Proteins of Living Cells
  • Flexible—Different Isotopes of Heavy and/or Light Amino Acids for Arginine, Lysine, Leucine and Proline Enable the Quantitation of Peptides Derived from MS-Grade Proteases
  • Multiplex Capabilities—Several Alternative Isotopes of Arginine and Lysine are Available that Allow the Analysis of Multiple Treatment Conditions in Each Experiment
  • High-Quality Supplements—Heavy Amino Acids with>99% Isotope Purity


This is a lyophilized preparation of L-Lysine-2HCl, sufficient for 10 SILAC experiments.


Stable isotope labeling with amino acids in cell culture (SILAC) is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of "heavy" 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.


Thermo Scientific Heavy and Light Amino Acids for SILAC are used together with specialized cell culture media that are deficient in essential amino acids. Heavy and light L-lysine and L-arginine are the most common amino acids used for SILAC analysis of tryptic peptides. Up to three different experimental conditions can be readily analyzed with different isotopes of lysine and arginine. For lysine three-plex experiments, 4,4,5,5-D4 L-lysine and 13C6 15N2 L-lysine are used to generate peptides with 4- and 8-Da mass shifts, respectively, compared to peptides generated with light lysine. For arginine three-plex experiments, 13C6 L-arginine and 13C6 15N4 L-arginine are used to generate peptides with 6- and 10-Da mass shifts, respectively, compared to peptides generated with light arginine. L-leucine is another amino acid commonly used for SILAC labeling, because it is one of the most common amino acids found in protein sequences. Proline is a non-essential amino acid that is sometimes added to SILAC media to prevent the metabolic conversion of heavy arginine to heavy proline in mammalian cell lines with high arginine dehydrogenase activity.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₆H₁₆Cl₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00066389
CAS Number: 657-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR