Leupeptin Protease Inhibitor
Supplier: Thermo Scientific
78435
CAPI78435EA 922.02 CAD
CAPI78435
Leupeptin Protease Inhibitor
Proteins and Peptides
Leupeptin Protease Inhibitor reversably inhibits serine and cysteine proteases.
  • High-quality reagents in convenient and affordable package sizes
  • Purchase individually to prepare customized protease inhibitor cocktails
  • Reliable supplement
Protease Inhibitors are small packages of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.
Caution: For Research Use Only. Not for use in diagnostic procedures.