Biodyne™ A Nylon Membrane, 0.45 µm, Thermo Scientific
Supplier: Thermo Scientific
77015
CAPI77015PK 1257.45 CAD
CAPI77015
Biodyne™ A Nylon Membrane, 0.45 µm, Thermo Scientific
Transfer Membranes
Thermo Scientific Biodyne A Nylon Membranes are sheets of unmodified nylon that provide excellent binding characteristics for nucleic acid transfer, blotting and detection methods.
  • Versatile and durable—achieve strong DNA binding by baking or UV-crosslinking without damaging the membrane
  • Easy to use—pre-cut 8 x 12 cm sheets are the ideal size for standard mini-gel transfers and microplate dot-blots
  • High signal-to-noise ratio—lower background than nitrocellulose and most other nylon transfer membranes
Biodyne A contains both negatively and positivelyMore Product Information