Pierce™ Protein IgG Plus Orientation Kit, Thermo Scientific
Supplier: Thermo Scientific
44893 44990
CAPI44893EA 1632.35 CAD
CAPI44893 CAPI44990
Pierce™ Protein IgG Plus Orientation Kit, Thermo Scientific
Protein Purification Protein Purification Kits
Pierce™ Protein IgG Plus Orientation Kits uses a crosslinker to covalently attach antibody that is bound to Protein A (or Protein G) agarose resin, thereby creating a reusable affinity column for purification of antigens.
  • Contaminant free antigen purification and less antibody leaching
  • Covalent, leak resistant antibody immobilization
  • Gentle binding and crosslinking conditions
  • Less antibody leaching
  • Crosslinked 6% beaded agarose with recombinant Protein A (or Protein G) thus making it high quality resin
  • Oriented immobilization that is able to capture passing antigen in the mobile phase without steric hindrance
Delivery information: Kit includes Recombinant Protein A [or Protein G] columns, 2 mL (2 columns), Disuccinimidyl suberate [DSS] (2 x 13 mg), PBS pack (makes 500 mL), Blocking Buffer (6 mL), IgG Elution Buffer (15 mL), Antibody Binding/Wash Buffer (120 mL), and column accessories.
Order Now
SPECIFICATIONS
Environmentally Preferable