Pierce™ Fast Western Blot Kits, SuperSignal™ West Dura, Mouse, Thermo Scientific
Supplier: Thermo Scientific
35070 35071
CAPI35070EA 1036.91 CAD
CAPI35070 CAPI35071
Pierce™ Fast Western Blot Kits, SuperSignal™ West Dura, Mouse, Thermo Scientific
Blotting Kits
The Thermo Scientific Pierce Fast Western Blot Kit, SuperSignal West Dura Substrate provides reagents and protocol for reliable and rapid, 1-hour chemiluminescent Western blotting at femtogram levels of sensitivity.
  • Fast—femtogram levels of detection in about 1 hour
  • Convenient—no expensive hardware or vacuum required; no clogging issues
  • Simple—optimized protocol makes Western blot analysis easier than ever
  • Economical—no expensive consumables or extra equipment
  • Excellent stability—kit is stable for 1 year stored at 4 °C
  • Easy—complete kits contain all
More Product Information