Pierce ProFound™ Biotinylated-Protein Interaction Pull Down Kit, Thermo Scientific
Supplier: Thermo Scientific
21115
CAPI21115EA 1442.44 CAD
CAPI21115
Pierce ProFound™ Biotinylated-Protein Interaction Pull Down Kit, Thermo Scientific
Assays Protein Assays
Pierce™ Biotinylated Protein Interaction Pull-Down Kit contains the necessary components to capture and purify interactors of a biotin-labeled protein or ligand.
  • Provides a complete easy-to-use strategy for discovering protein:protein interactions
  • Uses common laboratory equipment and reagents
  • Adaptable to single- or multiple-sample demands
  • Flexible pull-down format uses spin cups for easy and efficient manipulation of streptavidin agarose beads
Delivery information: Kit includes streptavidin Agarose resin (1.5 mL) settled gel; Lysis Buffer (250 mL); BupH Tris Buffered Saline, 1 pack (500 mL); Wash Buffer (Acetate, pH 5), (100 mL); Elution Buffer (pH 2.8), (50 mL); Spin Columns Accessory Pack, 27 columns; Collection tubes and caps (200x 2 mL tubes).