EDTA disodium salt dihydrate, Pierce™

Supplier: Thermo Scientific

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

17892
CAPI17892EA 262.06 CAD
CAPI17892
EDTA disodium salt dihydrate, Pierce™
EDTA disodium salt dihydrate

Thermo Scientific Pierce EDTA is useful as a chelator of alkaline earth metals, as well as iron, copper and zinc in a variety of laboratory methods.


  • Economical
  • Ready to use
  • Water-soluble


EDTA is especially useful for its ability to sequester metal ions to reduce their reactivity in solutions.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255
MDL Number: MFCD00150037
CAS Number: 6381-92-6
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR