Pierce™ Protein A, G, A/G Coated Plates Coated Plates, White, 96-Well, Thermo Scientific
Supplier: Thermo Scientific
15132 15133 15138 15130 15131 15154 15155 15156 15157
CAPI15132EA 659.2 CAD
CAPI15132 CAPI15133 CAPI15138 CAPI15130 CAPI15131 CAPI15154 CAPI15155 CAPI15156 CAPI15157
Pierce™ Protein A, G, A/G Coated Plates Coated Plates, White, 96-Well, Thermo Scientific
Microplates Immunology Microplates and Strips
Pierce™ Protein A, G and A/G coated plates provide an alternative to direct, passive adsorption methods for immobilising antibodies for ELISA and other plate-based assay techniques. Available in clear for colorimetric assays, white for chemiluminescent assays, and black for fluorescent assays.
  • Retain antibody activity, which can be lost when antibodies are immobilised by passive adsorption
  • Orient antibodies for maximum antigen-binding capacity
  • Immobilise antibodies for plate assays without prior purification
  • Ensure minimal variation (<5% well-to-well)
More Product Information