Protegrity® CP Sterile Latex Gloves with Nitrile, Cardinal
Supplier: Cardinal Health
2Y72N1 2Y72N2 2Y72N6 2Y72N7 2Y72N4
CAPH2Y72N1CS 587.45 CAD
CAPH2Y72N1 CAPH2Y72N2 CAPH2Y72N6 CAPH2Y72N7 PH2Y72N0 PH2Y72N4
Protegrity® CP Sterile Latex Gloves with Nitrile, Cardinal
Gloves
Triple-layer, powder-free, clean process gloves.
  • Triple layer, powder-free
  • Comfort of natural rubber latex with the strength of nitrile for exceptional grip and tactile properties
  • Hand specific, beaded cuff, bisque finish
  • Sterilized by gamma irradiation to a Sterility Assurance Level 10⁻⁶
Unique bonding technology integrates the comfort of natural rubber latex with the strength of nitrile for an exceptional grip, outstanding tactile properties, and enhanced comfort. The natural rubber latex outer layer provides elasticity and tensile strength.