CP100™ BT Sterile Latex Hand-Specific Gloves, Cardinal
Supplier: Cardinal Health
2Y5000
CAPH2Y5000CS 619.14 CAD
CAPH2Y5000
CP100™ BT Sterile Latex Hand-Specific Gloves, Cardinal
Gloves
Sterile, clean-processed gloves feature a comfortable flex-and-stretch design and low particulate levels for minimized contamination of controlled environments.
  • Ambidextrous, beaded cuff
  • Fully textured palm and fingers provide a secure grip
  • Clean processed, powder free
The antislip, textured finish enhances the grip for better handling. Tapered fingers help reduce fatigue while providing increased tactile sensitivity. Beaded cuff with reinforced ribs prevents constriction and reduces rolldown. Hand-specific. Powder-free. Color: natural white.