tert-Butyl methyl ether, anhydrous ≥99.8%, DriSolv®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether

Packed in septum-sealed bottle.

MX0824-7
CAMX0824-7EA 106 CAD
CAMX0824-7
tert-Butyl methyl ether, anhydrous ≥99.8%, DriSolv®, Supelco®
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
Boiling Pt: 55.2 °C (1013 hPa)
Melting Pt: –108.6 °C
Density: 0.7405 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.8% min
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Residue after evaporation 5ppm max
Water (H2O) 50ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR