Mouse Tissue, Rockland Immunochemicals
Supplier: Rockland Immunochemical
MS-T001 MS-T002 MS-T004 MS-T010 MS-T011 MS-T012 MS-T013 MS-T014 MS-T016 MS-T017 MS-T020 MS-T021 MS-T022 MS-T023 MS-T028 MS-T029 MS-T030 MS-T031 MS-T032 MS-T033 MS-T034 MS-T035 MS-T036
CAMS-T001EA 95.01 CAD
CAMS-T001 CAMS-T002 CAMS-T004 CAMS-T010 CAMS-T011 CAMS-T012 CAMS-T013 CAMS-T014 CAMS-T016 CAMS-T017 CAMS-T020 CAMS-T021 CAMS-T022 CAMS-T023 CAMS-T028 CAMS-T029 CAMS-T030 CAMS-T031 CAMS-T032 CAMS-T033 CAMS-T034 CAMS-T035 CAMS-T036
Mouse Tissue, Rockland Immunochemicals
Specimens Preserved Specimens Preserved Animal Materials
Use for DNA and RNA isolation.
  • Collected from antibiotic-free animals
  • DNA/RNA isolation
All tissues are collected from normal, healthy, mature animals that have been maintained on an antibiotic-free diet. Tissues are stored at −20°C or colder prior to use.
Ordering information: For specific breed, sex, or weight requirements, please contact your Avantor representative.