Potassium chloride saturated aqueous solution, StandARd®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Potassiumchloride

Management of Change (MOC) category = N

MACRH300-01 MACRH300-05
CAMKH30001EA 105.73 CAD
CAMKH30001 CAMKH30005
Potassium chloride saturated aqueous solution, StandARd®, Macron Fine Chemicals™
Potassium chloride

For refilling calomel reference electrodes.

Formula: KCl
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Density (g/mL) at 20°C 1.162 - 1.172
Identification Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR