Potassium nitrate 99.0-100.5%, fine crystals, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

MACR6715-06 MACR6715-04 MACR6715-20
CAMK671506EA 613.29 CAD
CAMK671506 CAMK671504 CAMK671520
Potassium nitrate 99.0-100.5%, fine crystals, Macron Fine Chemicals™
Potassium nitrate

Fine crystals.

Formula: KNO₃
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (KNO₃) 99.0 - 100.5 %
Solubility Passes Test
Loss on Drying at 105°C ≤1.0 %
Reaction Passes Test
Identification A Passes Test
Identification B Passes Test
Identification for Sodium Passes Test
Sodium (Na), For Information Only
Chlorate Passes Test
Chloride (Cl) ≤0.058 %
Arsenic (As) ≤0.0003 %
Heavy Metals (as Pb) ≤0.0020 %
Lead (Pb) ≤0.0010 %
Appearance (white to off-white crystals)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR