Hydrogen peroxide solution_SC 3% KMnO4 titration, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


M.O.C. Category N

MACR5241-05 MACR5241-08 MACR5241-45 MACR5241-04
CAMK524105CS 1237.01 CAD
CAMK524105 CAMK524108 CAMK524145 CAMK524104
Hydrogen peroxide solution_SC 3% KMnO4 titration, AR®, Macron Fine Chemicals™
Hydrogen peroxide

Clear, colorless liquid.

Formula: H₂O₂
MW: 34.01
Boiling Pt: 150.2
Melting Pt: -0.41
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
Merck Index: 14,04798

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (H₂O₂) (by KMnO₄ titrn) 3.10 - 3.30 %
Arsenic (As) ≤0.00005 %
Barium (Ba) Passes Test
Chloride (Cl) ≤0.001 %
Free Acid (as H₂SO₄) ≤0.010 %
Heavy Metals (as Pb) ≤0.0005 %
Iron (Fe) ≤0.0005 %
Nitrogen Compounds (as N) ≤0.005 %
Phosphate (PO₄) ≤0.003 %
Residue after Evaporation ≤0.020 %
Sulfate (SO₄) ≤0.010 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR