Magnesium perchlorate, AR® ACS desiccant, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Anhydrone

Management of Change (MOC) category = N

MACR5039-04
CAMK503904CS 13717.54 CAD
CAMK503904
Magnesium perchlorate, AR® ACS desiccant, Macron Fine Chemicals™
Magnesium perchlorate

For drying.

Formula: Cl₂MgO₈
MW: 223.21 g/mol
Melting Pt: 251
Density: 2.60 g/cm³
MDL Number: MFCD00011102
CAS Number: 10034-81-8
EINECS: 233-108-3
UN: 1475
ADR: 5.1 II
Merck Index: 13,05702

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Loss on Drying at 190°C ≤8 %
ACS - Suitability for Moisture Absorption Passes Test
ACS - Titrable Free Acid (meq/g) ≤0.005
ACS - Titrable Base (meq/g) ≤0.025
Appearance (white granular or flaky powder)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR