Ethyl acetate ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Acetic acid ethyl ester

Management of Change (MOC) category = N

4992-08 MACR4992-04 MACR4992-06 MACR4992-19 MACR4992-26
MK499208CS 2394.52 CAD
MK499208 CAMK499204 CAMK499206 CAMK499219 CAMK4992-26
Ethyl acetate ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃COOC₂H₅) (by GC, corrected for water) ≥99.5 %
Appearance (clear, colorless liquid) Passes Test
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.003 %
ACS - Solubility in H₂O Passes Test
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Titrable Acid (meq/g) ≤0.0009
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR