Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dry basis), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

MACR1806-10 MACR1806-11 MACR1806-57
CAMK180610CS 5082.01 CAD
CAMK180610 CAMK180611 CAMK180657 CAMK180688
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dry basis), AR® ACS, Macron Fine Chemicals™
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula: NH₂C(CH₂OH)₃
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 169…172 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (NH₂C(CH₂OH)₃) (dried basis) 99.8 - 100.1 %
ACS - Insoluble Matter ≤0.005 %
ACS - Loss on Drying at 105°C ≤1.0 %
ACS - pH of 0.05M Solution at 25°C 10.3 - 10.5
ACS - Water (H₂O)(by Karl Fischer titrn) ≤2 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤5 ppm
ACS - Absorbance Passes Test
Appearance (white crystalline granules or powder)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR