Potassium chlorate ≥99.0% (by KMnO₄ titration), granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

MACR6834-04
CAMK6834-04CS 4719.7 CAD
CAMK6834-04 CAMK-6834-04
Potassium chlorate ≥99.0% (by KMnO₄ titration), granular, AR® ACS, Macron Fine Chemicals™
Potassium chlorate
Formula: KClO₃
MW: 122.55 g/mol
Boiling Pt: 400
Melting Pt: 356
Density: 2.30 g/cm³
MDL Number: MFCD00011361
CAS Number: 3811-04-9
UN: 1485
ADR: 5.1 II
Merck Index: 13,07703

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (KClO₃) (by KMnO₄ titrn) ≥99.0 %
ACS - Bromate (BrO₃) ≤0.015 %
ACS - Calcium (Ca) ≤0.002 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤3 ppm
ACS - Magnesium (Mg) ≤0.002 %
ACS - Sodium (Na) ≤0.01 %
ACS - Sulfate (SO₄) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR