Phenol red sodium salt, powder, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide

Management of Change (MOC) category = N

BAKRT265-01 BAKRT265-03 BAKRT265-08
CAJTT265-1EA 497.16 CAD
CAJTT265-1 CAJTT265-3 CAJTT265-8
Phenol red sodium salt, powder, BAKER ANALYZED® ACS
Phenol red sodium salt

Powder. Lot analysis on label.

Formula: C₁₉H₁₃NaO₅S
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS Number: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Clarity of Solution Passes Test
Visual Transition Interval - pH…8.2 (Red) Passes Test
Visual Transition Interval - pH…6.8 (Yellow) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR