N-Methyl-2-pyrrolidone (NMP) ≥99% (by GC, corrected for water content), BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

Management of Change (MOC) category = N

BAKRR053-07 BAKRR053-09
CAJTR053-7CS 3841.03 CAD
CAJTR053-7 CAJTR053-9
N-Methyl-2-pyrrolidone (NMP) ≥99% (by GC, corrected for water content), BAKER ANALYZED®
N-Methyl-2-pyrrolidone (NMP)

Lot analysis on label.

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₅H₉NO) (by GC, corrected for water) ≥99 %
Color (APHA) ≤50
Density (g/mL) at 25°C 1.024 - 1.028
Water (H₂O)(by Karl Fischer titrn) ≤0.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR