J.T.Baker® DILUT-IT™, Sodium Phosphate, Dissolution Media Concentrate

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

BAKRD021-00 BAKRD021-02
CAJTD021-0CS 1999.47 CAD
CAJTD021-0 CAJTD021-2 CAJTD021-4
J.T.Baker® DILUT-IT™, Sodium Phosphate, Dissolution Media Concentrate
Buffers Biological Buffers

  • pH at 25 °C at 26X Dilution: 6.75 to 6.85
  • Store at room temperature

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR