Toluene 99,7 GC, corrected for water content, BAKER ANALYZED® HPLC HPLC/UHPLC, Spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

M.O.C. Category N

BAKR9351-02 BAKR9351-03 BAKR9351-33
CAJT9351-2CS 1170.65 CAD
CAJT9351-2 CAJT9351-3 CAJT9351-33
Toluene 99,7 GC, corrected for water content, BAKER ANALYZED® HPLC HPLC/UHPLC, Spectrophotometry, J.T.Baker®
Toluene

Packaged under nitrogen. For use in high performance liquid chromatography. UV cutoff 285nm. Lot analysis on label.


  • UV cutoff 285 nm

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (by GC, corrected for water) ≥99.7 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 288 nm ≤0.40
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤285
Residue after Evaporation ≤3.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR