Cyclohexane 99 GC, corrected for water content, BAKER ANANYZED™ ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

M.O.C. Category N

BAKR9206-01 BAKR9206-33 BAKR9206-05 9206-22
CAJT9206-1CS 2227.97 CAD
CAJT9206-1 CAJT9206-22 CAJT9206-33 CAJT9206-5 JT9206-22
Cyclohexane 99 GC, corrected for water content, BAKER ANANYZED™ ACS
Cyclohexane

Lot analysis on label.

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81
Melting Pt: 7
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₂) (by GC, corrected for water) ≥99.0 %
Appearance Passes Test
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.02 %
Meets Reagent Specifications for testing USP/NF monographs

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR