J.T.Baker® BAKERBOND Speedisk® Extraction Disks and Stations
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MACR8064-06 BAKR8094-06 BAKR8095-06 BAKR8055-06 BAKR8055-07 BAKR8056-06 BAKR8057-06 BAKR8058-06 BAKR8060-06 BAKR8061-06 BAKR8068-06 BAKR8072-06 BAKR8072-07 BAKR8082-06
CAJT8064-6PK 1359.8 CAD
CAJT8064-6 CAJT8094-6 CAJT8095-6 CAJT80556 CAJT80557 CAJT80566 CAJT80576 CAJT80586 CAJT80606 CAJT80616 CAJT80686 CAJT80687 CAJT80726 CAJT80727 CAJT80826
J.T.Baker® BAKERBOND Speedisk® Extraction Disks and Stations
Filters Filter Discs
Disks are preassembled for use in preparing aqueous samples for analysis.
Laminar configuration provides filtration capacity and inlet characteristics that maximize access of analyte molecules to the micro-particulate sorbent. Disk design resists clogging and ensures high throughput rates even when samples contain solids.
Order Now
SPECIFICATIONS
Environmentally Preferable