Perchloric acid 0.1 N in glacial acetic acid, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

BAKR5624-02
CAJT5624-2CS 3491.45 CAD
CAJT5624-2
Perchloric acid 0.1 N in glacial acetic acid, BAKER ANALYZED® volumetric solution, J.T.Baker®
Perchloric acid

The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.

Formula: HClO₄
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality, For Information Only
Standardization at 25°C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR