Sodium hydroxide concentrate for dilution to 0.1 N aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda

M.O.C. Category N

BAKR4687-01
CAJT4687-1CS 896.48 CAD
CAJT4687-1
Sodium hydroxide concentrate for dilution to 0.1 N aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sodium hydroxide

(1/10 equiv.=4.000 g NaOH)

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.327 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

For laboratory research or manufacturing use.
Normality (by titrimetry) Passes Test
(Makes 0.1N solution after dilution to 1000 ml)
Low in carbonate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR