HEPES sodium salt ≥99.0% (dried basis), Ultrapure Bioreagent for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonate, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonate, Sodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonate, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium salt, 4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt, HEPES sodium, Sodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulfonate, HEPES Na

Management of Change (MOC) category = N

BAKR4153-05
CAJT4153-1EA 1211.4 CAD
CAJT4153-1 CAJT4153-5 CAJT4153-6 JT4153-1
HEPES sodium salt ≥99.0% (dried basis), Ultrapure Bioreagent for molecular biology, J.T.Baker®
HEPES sodium salt

Ultrapure Bioreagent - A line of high purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography. Where applicable, tests include DNase, RNase, Protease, heavy metals and insoluble matter.

Formula: C₈H₁₇N₂NaO₄S
MW: 260.29 g/mol
Melting Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS Number: 75277-39-3
EINECS: 278-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116e9b4f3c17d013ff2b7c9848c219943a725b3c&vid=8c800de68c4434c3c9d77107ff6779ef516bd851&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₈H₁₇N₂NaO₄S) (dried basis) ≥99.0 %
Appearance Passes Test
Solubility Passes Test
Loss on Drying at 80°C ≤3.0 %
Insoluble Matter ≤1.0 %
Heavy Metals (as Pb) ≤1 ppm
Identification (Sodium) Passes Test
pH of 5% Solution at 25°C 9.5 - 11.5
pK^a at 20°C, For Information Only
QC Supervisor or equivalent to check supplier's COA to ensure proper chemical form. Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR